Learn More
Honeywell Burdick & Jackson™ SSPDS Accessory, MNPT Hose
For use with SSPDS
Manufacturer:  Honeywell Burdick & Jackson™ CA5H4NPT
 View more versions of this product
Catalog No. 60-029-34
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
  • Pre-cleaned PTFE hose
  • encased in a flexible stainless steel outer braid
Specifications
Specifications
5 ft.
1/4 in.