Learn More
Honeywell Burdick & Jackson™ SSPDS Accessory, STB Hose
For use with SSPDS
Manufacturer:  Honeywell Burdick & Jackson™ CA10H8STB
 View more versions of this product
Catalog No. 60-029-19
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
  • Pre-cleaned PTFE hose
  • encased in a flexible stainless steel outer braid
Specifications
Specifications
10 ft.
1/2 in.