Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SSR2 Mouse anti-Human, Alexa Fluor™ 350, Clone: 31G6, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP242647AF350

Catalog No. NP242647A30


Add to cart

Description

Description

SSR2 Monoclonal antibody specifically detects SSR2 in Human samples. It is validated for Western Blot, ELISA.
Specifications

Specifications

SSR2
31G6
Western Blot, ELISA
Conjugated, Purified
SSR2
Recombinant human SSR2 (18-149aa) purified from E. coli
Protein A purified
Store at 4°C in the dark.
Monoclonal
Human
ELISA, Western Blot
Alexa Fluor™ 350
50 mM sodium borate with 0.05% sodium azide
DKFZp686F19123, Signal sequence receptor subunit beta, signal sequence receptor, beta (translocon-associated protein beta), SSR-beta, TLAP, translocon-associated protein beta, translocon-associated protein subunit beta, TRAP-beta, TRAPBTRAP-BETA
Mouse
IgG1 Kappa
100 μL
Primary
6746.0
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit