Learn More
SSRP1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 156961AP
Catalog No. 50-173-0413
Add to cart
Description
Description
This gene encodes a protein that belongs to the microtubule-associated protein family. The proteins of this family are thought to be involved in microtubule assembly, which is an essential step in neurogenesis. The product of this gene is a precursor polypeptide that presumably undergoes proteolytic processing to generate the final MAP1B heavy chain and LC1 light chain. Gene knockout studies of the mouse microtubule-associated protein 1B gene suggested an important role in development and function of the nervous system.
Specifications
Specifications
SSRP1
0.17 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q04931, Q08943, Q08945
FACT, FACT 80 kDa subunit, FACT complex subunit SSRP1, FACT80, FACTp80, hSSRP1, SSRP1, T160
Rabbit
IgG
150 μL
-20°C
Primary
20833, 6749, 81785
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Unconjugated
SSRP1
Liquid
SSRP1
SSRP1 Fusion Protein Ag8318
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat