Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SSSCA1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5112726
Description
SSSCA1 Polyclonal Antibody for Western Blot, IHC (P), ELISA
Might play a role in mitosis. Antigenic molecule. Could be a centromere-associated protein. May induce anti-centromere antibodies.
Specifications
SSSCA1 | |
Polyclonal | |
Unconjugated | |
ZNRD2 | |
1500016H19Rik; Autoantigen p27; autoantigen p27 homolog; C184l; centromeric autoantigen (27kD); p27; Protein C184L; Protein ZNRD2; Sjoegren syndrome/scleroderma autoantigen 1; Sjoegren syndrome/scleroderma autoantigen 1 homolog; Sjogren syndrome/scleroderma autoantigen 1; Sjogren syndrome/scleroderma autoantigen 1 homolog; Sjogren's syndrome/scleroderma autoantigen 1; Sjogren's syndrome/scleroderma autoantigen 1 homolog; Sjogren's syndrome/scleroderma autoantigen 1 homolog (human); SSSCA1; Zinc ribbon domain-containing protein 2; Znrd2 | |
Rabbit | |
Protein G | |
RUO | |
10534 | |
-20°C or -80°C if preferred | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Western Blot | |
4 mg/mL | |
PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
O60232 | |
ZNRD2 | |
Recombinant Human Sjoegren syndrome/scleroderma autoantigen 1 protein (2-199AA). | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction