Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SSSCA1 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA548613
Description
Might play a role in mitosis. Antigenic molecule. Could be a centromere-associated protein. May induce anti-centromere antibodies.Specifications
SSSCA1 | |
Polyclonal | |
PBS with 0.09% sodium azide | |
O60232, P56873 | |
SSSCA1 | |
KLH conjugated synthetic peptide between 9-37 amino acids from the N-terminal region of human SSSCA1. | |
400 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
SSSCA1 | |
Sjoegren syndrome/scleroderma autoantigen 1, Autoantigen p27, SSSCA1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
10534, 56390 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction