Learn More
MBL International Corporation SSTR1 Antibody (N-Terminus) Polyclonal Antibody (US Only)
Supplier:  MBL International Corporation MC994
Encompass_Preferred
Clonality:Polyclonal; Application:IHC; Conjugate:unconjugated; Species Reactivity:Human, Monkey, Mouse, Bovine, Dog, Hamster, Horse, Pig, Rabbit; Size:50 µg; Target:SSTR1; SSTR1 Primary Antibody of 50 µg. For sale In the US only. Somatostatins are peptide hormones that regulate diverse cellular functions such as neurotransmission, cell proliferation, and endocrine signaling as well as inhibiting the release of many hormones and other secretory proteins. Somatostatin has two active forms of 14 and 28 amino acids. The protein encoded by this gene is a member of the superfamily of somatostatin receptors having seven transmembrane segments. Somatostatin receptors form homodimers and heterodimers with other members of the superfamily as well as with other G-protein coupled receptors and receptor tyrosine kinases. This somatostatin receptor has greater affinity for somatostatin-14 than -28.
Catalog No. 50-495-30