Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SSTR3 Mouse anti-Human, Rat, Clone: 7H8E5, Abnova™

Mouse monoclonal antibody raised against partial recombinant human SSTR3.

Manufacturer:  Abnova Corporation MAB12261

Catalog No. 89-934-969


Add to cart

Description

Description

Somatostatin acts at many sites to inhibit the release of many hormones and other secretory proteins. The biological effects of somatostatin are probably mediated by a family of G protein-coupled receptors that are expressed in a tissue-specific manner. SSTR3 is a member of the superfamily of receptors having seven transmembrane segments and is expressed in highest levels in brain and pancreatic islets. SSTR3 is functionally coupled to adenylyl cyclase. [provided by RefSeq]

Specifications

Specifications

SSTR3
7H8E5
Mouse monoclonal antibody raised against partial recombinant human SSTR3.
In PBS (0.05% sodium azide)
SSTR3
Recombinant protein corresponding to amino acids 1-43 of human SSTR3.
100 ug
Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Monoclonal
Human, Rat
ELISA, Flow Cytometry, Western Blot
Unconjugated
ELISA (1:10000) Flow Cytometry (1:200-1:400) Western Blot (1:500-1:2000) The optimal working dilution should be determined by the end user.
Liquid
Mouse
IgG1
RUO
Primary
6753
SDS
Documents
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit