Learn More
Invitrogen™ SSTR5 Polyclonal Antibody

Description
This antibody is predicted to detect chimp Somatostain receptor 5 based on 94% sequence homology. Suggested positive control: human pancreas cell lysate, human pancreas protein.
Specifications
Specifications
| Antigen | SSTR5 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.2% gelatin and 0.05% sodium azide |
| Gene | Sstr5 |
| Gene Accession No. | P35346 |
| Gene Alias | Smstr5; Somato; somatostatin receptor; somatostatin receptor 5; Somatostatin receptor subtype 5; somatostatin receptor type 5; SS5R; SS-5-R; SS5-R; sst5; Sstr5 |
| Gene Symbols | Sstr5 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.