Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ssu72 Mouse anti-Human, Clone: 66D10, Novus Biologicals™
Mouse Monoclonal Antibody
$337.00 - $482.50
Specifications
Antigen | Ssu72 |
---|---|
Clone | 66D10 |
Concentration | 1 mg/mL |
Dilution | Western Blot 1:1000, ELISA 1:100 - 1:2000 |
Applications | Western Blot, ELISA |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB6023650UL
|
Novus Biologicals
NBP26023650UL |
50 μL |
Each for $337.00
|
|
|||||
NB60236100U
|
Novus Biologicals
NBP260236100UL |
100 μL |
Each for $482.50
|
|
|||||
Description
Ssu72 Monoclonal specifically detects Ssu72 in Human samples. It is validated for Western Blot, ELISA.Specifications
Ssu72 | |
1 mg/mL | |
Western Blot, ELISA | |
Unconjugated | |
Mouse | |
Human | |
CTD phosphatase SSU72, EC 3.1.3.16, FLJ13048, HSPC182, PNAS-120, RNA polymerase II subunit A C-terminal domain phosphatase SSU72, SSU72 RNA polymerase II CTD phosphatase homolog (S. cerevisiae), Ssu72 RNA polymerase II CTD phosphatase homolog (yeast) | |
SSU72 | |
IgG2b κ | |
Protein A purified |
66D10 | |
Western Blot 1:1000, ELISA 1:100 - 1:2000 | |
Monoclonal | |
Purified | |
RUO | |
PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide | |
29101 | |
Recombinant human SSU72 (1-194aa) purified from E. coli | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
RUO
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title