Learn More
SSU72 Mouse anti-Human, Clone: OTI2E2, lyophilized, TrueMAB™ Non-distribution product as customer accommodation.
Mouse Monoclonal Antibody
Manufacturer:  OriGeneSupplier Diversity Partner CF809191
Catalog No. 50-166-9330
Add to cart
Description
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Human Ssu72 is a highly conserved homologue of yeast Ssu72, a CTD phosphatase and a component of the polyadenylation/ termination machinery. The human Ssu72 is located on Chromosome 1p36 and seems to represent a single gene. It was identified as a pRb binding factor and was also shown to interact with TFIIB and yeast PTA1.
Specifications
Specifications
SSU72
Monoclonal
Unconjugated
SSU72
Lyophilized
SSU72
Full length human recombit protein of human SSU72 produced in E.coli.
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Immunohistochemistry (Paraffin), Western Blot
OTI2E2
PBS with 8% trehalose and no preservative; pH 7.3
Q9NP77
HSPC182, PNAS-120
Mouse
IgG1
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
29101