Learn More
SSU72 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
SDP
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group Inc 154341AP150UL
Encompass_Preferred
 View more versions of this product
Catalog No. 50-173-0224
Add to cart
Description
Description
SSU72 is a highly conserved homologue of yeast Ssu72, a CTD phosphatase and a component of the polyadenylation/ termination machinery. The human Ssu72 is located on Chromosome 1p36 and seems to represent a single gene. It was identified as a pRb binding factor and was also shown to interact with TFIIB and yeast PTA1. The ABs recognize a single band of approximately 30 Kda on immunoblots of various human cell lines but do not work well on mouse tissues.
Specifications
Specifications
SSU72
Polyclonal
Unconjugated
SSU72
CTD phosphatase SSU72, HSPC182, PNAS 120, SSU72
Rabbit
Antigen Affinity Chromatography
RUO
29101, 298681, 68991
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.13 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q4KLK9, Q9CY97, Q9NP77
SSU72
SSU72 Fusion Protein Ag7699
150 μL
Primary
Human, Mouse, Rat
Antibody
IgG