Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SSX1 Mouse anti-Human, Clone: OTI1D10, liquid, TrueMAB™
Mouse Monoclonal Antibody
Supplier: OriGene TA502523
Description
Could act as a modulator of transcription.Specifications
SSX1 | |
Monoclonal | |
0.59 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
Q16384 | |
SSX1 | |
Full length human recombit protein of human SSX1 produced in E.coli. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot | |
OTI1D10 | |
Unconjugated | |
SSX1 | |
CT5.1, SSRC | |
Mouse | |
Affinity Chromatography | |
RUO | |
6756 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction