Learn More
Invitrogen™ SSX5 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5117104
Description
Positive Samples: Rat liver Immunogen sequence: MNGDDAFVRR PRVGSQIPEK MQKHPWRQVC DRGIHLVNLS PFWKVGREPA SSIKALLCGR GEARAFDDIA KYFSEKEWEK MKASEKIIYV YMKRKYEAMT KLGFKATLPP FMRNKRVADF QGNDFDNDPN RGNQVEHPQM TFGRLQGIFP KITPEKPAEE GNDSKGVPEA SGPQNNGKQL RPSGKLNTSE KVNKTSGPKR GKHAWTHRVR ERKQLVIYEE ISDPQEDDE.
The product of this gene belongs to the family of highly homologous synovial sarcoma X (SSX) breakpoint proteins. These proteins may function as transcriptional repressors. They are also capable of eliciting spontaneous humoral and cellular immune responses in cancer patients, and are potentially useful targets in cancer vaccine-based immunotherapy. While some of the related SSX genes are involved in t(X; 18)(p11.2; q11.2) translocations that are characteristically found in all synovial sarcomas, this gene does not appear to be involved in such translocations. Two transcript variants encoding distinct isoforms have been identified for this gene.
Specifications
SSX5 | |
Polyclonal | |
Unconjugated | |
SSX5 | |
protein SSX5; SSX family member 5; SSX5; synovial sarcoma, X breakpoint 5 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
6758 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
1.1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O60225 | |
SSX5 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-229 of human SSX5 (NP_0662953). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.