Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ ST 045849

O-GlcNAc transferase (OGT) inhibitor

$215.50 - $908.50

Specifications

CAS 442665-87-4
Chemical Name or Material 2-[(4-Chlorophenyl)imino]tetrahydro-4-oxo-3-(2-tricyclo[3.3.1.13,7]dec-1-ylethyl)-2H-1,3-thiazine-6-carboxylic acid
Molecular Formula C23H27ClN2O3S
Purity 98%
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
67-751-0
View Documents
Tocris Bioscience™
6775/10
10 mg
Each for $215.50
Add to cart
 
67-755-0
View Documents
Tocris Bioscience™
6775/50
50 mg
Each for $908.50
Add to cart
 
Description

Description

  • O-GlcNAc transferase (OGT) inhibitor (IC50 = 53 μM). Reduces proliferation and viability of prostate cancer cells in vitro.
  • Depletes intracellular alanine and decreases glucose consumption by cancer cells.
  • Decreases markers of ER stress and apoptosis in pregnant diabetic mice and reduces neural tube defects in embryos from diabetic mice.
Specifications

Specifications

442665-87-4
2-[(4-Chlorophenyl)imino]tetrahydro-4-oxo-3-(2-tricyclo[3.3.1.13,7]dec-1-ylethyl)-2H-1,3-thiazine-6-carboxylic acid
C23H27ClN2O3S
98%
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit