Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ST 91, Tocris Bioscience™
Click to view available options
Quantity:
10 mg
50 mg
Description
α2-adrenoceptor agonist that does not cross the blood brain barrier. Displays ∽ 120-fold selectivity for α2 receptors over α1 receptors and acts predominantly at non-α2A-adrenoceptors which may be of the α2C subtype. Antinociceptive; increases tail-flick latencies in Sasco and Harlan rats.
Specifications
Quantity | 10 mg |
Formula Weight | 253.77 |
Percent Purity | >99% |
Chemical Name or Material | ST 91 |
Safety and Handling
Recommended Storage : Desiccate at +4°C