Learn More
Enzo Life SciencesSupplier Diversity Partner ST 2 polyclonal antibody (100 µg)  DFS Item
Manufacturer:  Enzo Life SciencesSupplier Diversity Partner ADICSA512E
Host: Rabbit. Immunogen: Synthetic peptide corresponding to the sequence near the N-terminus of mouse ST2. The sequence is common to all three ST2 isoforms. UniProt: P14719. Species Reactivity: Human, Mouse. Applications: IHC, WB. Recommended dilutions: Immunohistochemistry (2µg/ml). Western Blot (1-2µg/ml). Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Purity: Peptide affinity purified. Formulation: Liquid. In PBS containing 0.02% sodium azide.
Catalog No. 50-659-019