Learn More
ST2/IL-33R Rabbit anti-Human, Alexa Fluor 405, Clone: 2154B, R&D Systems™
Rabbit Monoclonal Antibody
Manufacturer:  R&D Systems FAB5232V100UG
Catalog No. FB5232V100U
Add to cart
This item is not returnable. View return policy
Description
Description
ST2/IL-33R Monoclonal specifically detects ST2/IL-33R in Human samples. It is validated for Flow Cytometry.
Specifications
Specifications
ST2/IL-33R
Monoclonal
Alexa Fluor 405
Supplied in a saline solution containing BSA and Sodium Azide.
DER4, DER4ST2growth stimulation-expressed, FIT-1, IL-1 R4, IL1R4, IL-1R4, IL-1RL1, IL-33 R, IL33R, IL-33R, interleukin 1 receptor-like 1, interleukin 1 receptor-related protein, interleukin-1 receptor-like 1, Ly84, MGC32623, Protein ST2, ST2L, ST2V, T1, T1homolog of mouse growth stimulation-expressed
S. frugiperda insect ovarian cell line Sf 21-derived recombinant human ST-2/ IL-33R Lys19-Phe328 Accession # Q01638
100 μg
Primary
Detects human ST-2/IL-33R in direct ELISAs.
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
IgG
Flow Cytometry
2154B
Flow Cytometry 0.25-1 ug/10^6 cells
Q01638
Rabbit
Protein A or G purified from cell culture supernatant
RUO
9173
Human
Purified