Learn More
ST2/IL-33R Rat anti-Mouse, R-PE, Clone: 245707, R&D Systems
Rat Monoclonal Antibody
Manufacturer:  R&D Systems FAB10041P100UG
Catalog No. FB10041P100
Add to cart
This item is not returnable. View return policy
Description
Description
ST2/IL-33R Monoclonal antibody specifically detects ST2/IL-33R in Mouse samples. It is validated for Flow Cytometry.
Specifications
Specifications
ST2/IL-33R
245707
Flow Cytometry 0.25-1 μg/106 cells
P14719
DER4, DER4ST2growth stimulation-expressed, FIT-1, IL-1 R4, IL1R4, IL-1R4, IL-1RL1, IL-33 R, IL33R, IL-33R, interleukin 1 receptor-like 1, interleukin 1 receptor-related protein, interleukin-1 receptor-like 1, Ly84, MGC32623, Protein ST2, ST2L, ST2V, T1, T1homolog of mouse growth stimulation-expressed
Rat
IgG2b
100 μg
Primary
9173.0
Mouse
Flow Cytometry
R-PE
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
Purified, Supernatant, Conjugated
Il1rl1
S. frugiperda insect ovarian cell line Sf 21-derived recombinant mouse ST2/IL-33 R, Ser27-Arg332, Accession No. P14719
Protein A or G purified from hybridoma culture supernatant
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied.
Monoclonal
Detects mouse ST2/IL-33 R in ELISAs.