Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human ST2/IL-33R Alexa Fluor™ 700-conjugated Antibody, R&D Systems™
Rabbit Monoclonal Antibody
Supplier: R&D Systems FAB10118N100UG
This item is not returnable.
View return policy
Description
ST2/IL-33R Monoclonal specifically detects ST2/IL-33R in Human samples. It is validated for Flow Cytometry.Specifications
ST2/IL-33R | |
Monoclonal | |
Alexa Fluor 700 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and sodium azide | |
DER4, DER4ST2growth stimulation-expressed, FIT-1, IL-1 R4, IL1R4, IL-1R4, IL-1RL1, IL-33 R, IL33R, IL-33R, interleukin 1 receptor-like 1, interleukin 1 receptor-related protein, interleukin-1 receptor-like 1, Ly84, MGC32623, Protein ST2, ST2L, ST2V, T1, T1homolog of mouse growth stimulation-expressed | |
S. frugiperda insect ovarian cell line Sf 21-derived recombinant human ST2/IL-33 R, Lys19-Phe328, Accession No. Q01638 | |
Primary | |
Detects human ST2/IL-33R in direct ELISAs. In Flow Cytometry, it detects human ST2/IL-33R-transfected HEK293 cells but not irrelevant or non-transfected cells. | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied. |
Flow Cytometry | |
2154E | |
Flow Cytometry 0.25-1 μg/106 cells | |
Q01638 | |
Rabbit | |
100 μg | |
9173 | |
Human | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction