Learn More
Invitrogen™ ST3GAL3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5110436
Description
Immunogen sequence: LHLLQWEEDS NSVVLSFDSA GQTLGSEYDR LGFLLNLDSK LPAELATKYA NFSEGACKPG YASALMTAIF PRFSKPAPMF LDDSFRKWAR IREFVPPFGI KGQDNLIKAI LSVTKEYRLT PALDSLRCRR CIIVGNGGVL ANKSLGSRID DYDIVVRLNS APVKGFEKDV GSKTTLRITY PEGAMQRPEQ YERDSLFVLA GFKWQDFKWL KYIVYKERVS ASDGFWKSVA TRVPKEPPEI RILNPYFIQE AAFTLIGLPF NNGLMGRGNI PTLGSVAVTM ALHGCDEVAV AGFGYDMSTP NAPLHYYETV RMAAIKESWT HNIQREKEFL RKLVKARVIT DLSSGI.
The protein encoded by this gene is a type II membrane protein that catalyzes the transfer of sialic acid from CMP-sialic acid to galactose-containing substrates. The encoded protein is normally found in the Golgi apparatus but can be proteolytically processed to a soluble form. This protein is a member of glycosyltransferase family 29. Multiple transcript variants encoding several different isoforms have been found for this gene.
Specifications
ST3GAL3 | |
Polyclonal | |
Unconjugated | |
ST3GAL3 | |
alpha 2,3-sialyltransferase III; alpha 2,3-ST 3; alpha-2,3-sialyltransferase II; beta-galactoside alpha-2,3-sialyltransferase 3; CMP-N-acetylneuraminate-beta-1,4-galactoside alpha-2,3-sialyltransferase; CMP-N-acetylneuraminate-beta-1,4-galactoside alpha-2,3-sialyltransferase soluble form; CMP-N-acetylneuraminate-beta-1,4-galactoside alpha-2,3-sialyltransferase; LOW QUALITY PROTEIN: CMP-N-acetylneuraminate-beta-1,4-galactoside alpha-2,3-sialyltransferase; EIEE15; gal beta-1,3(4) GlcNAc alpha-2,3 sialyltransferase; Gal beta-1,3(4)GlcNAc alpha-2,3 sialyltransferase; mental retardation, non-syndromic, autosomal recessive, 12; MRT12; N-acetyllacosaminide alpha 2,3-sialyltransferase; N-acetyllactosaminide alpha-2,3-sialyltransferase; sialyltransferase (N-acetyllacosaminide alpha 23-sialyltransferase); sialyltransferase 3; sialyltransferase 6; sialyltransferase 6 (N-acetyllacosaminide alpha 2,3-sialyltransferase); sialyltransferase 6(N-acetyllacosaminide alpha 23-sialyltransferase); sialyltransferase ST3Gal-III; Siat3; Siat6; ST3 beta-galactoside alpha-2,3-sialyltransferase 3; ST3Gal III; St3gal3; ST3GALII; ST3GalIII; ST3N | |
Rabbit | |
Affinity Chromatography | |
RUO | |
20441, 64445, 6487 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
0.52 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P97325, Q02734, Q11203 | |
ST3GAL3 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 30-375 of human ST3GAL3 (NP_006270.1). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.