Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ ST6 Gal Sialyltransferase 1/ST6GAL1/CD75 Antibody Pair [HRP]

Polyclonal/Monoclonal Antibody Pair

Manufacturer:  Novus Biologicals™ NBP2796565PLATES

 View more versions of this product

Catalog No. NB010352


Add to cart

Description

Description

This solid phase sandwich ELISA utilizes an antibody specific for Human ST6 Gal Sialyltransferase 1/ST6GAL1/CD75.
Specifications

Specifications

Sandwich ELISA
HRP
78.13 pg/mL
6480
RUO
ST6 Gal Sialyltransferase 1/ST6GAL1/CD75
Golgi Apparatus Markers
Human
ST6GAL1
1. Capture Antibody: Dilute to a working concentration of 2 μg/mL in PBS before coating. 2. Detection Antibody: Dilute to working concentration of 0.5 μg/mL in detection antibody dilution buffer before use. 3. Standard: Reconstitute with 1 ml detection antibody dilution buffer. After reconstitution, store at -20 to -80°C in a manual defrost freezer. A seven-point standard curve using 2-fold serial dilutions in sample dilution buffer, and a high standard of 5000 pg/mL is recommended.
This solid phase sandwich ELISA utilizes an antibody specific for Human ST6 Gal Sialyltransferase 1/ST6GAL1/CD75.
Antibody Pairs
Sandwich ELISA
  • Standard: Each vial contains 83 ng of recombinant Human ST6 Gal Sialyltransferase 1/ST6GAL1/CD75.
  • Capture Antibody: 1 mg/mL of rabbit anti-Human ST6 Gal Sialyltransferase 1/ST6GAL1/CD75 monoclonal antibody (in PBS, pH 7.4).
  • Detection Antibody: 0.2 mg/mL of rabbit anti-Human ST6 Gal Sialyltransferase 1/ST6GAL1/CD75 polyclonal antibody conjugated to horseradish-peroxidase (HRP) (in PBS, 50 % HRP-Protector, pH 7.4, store at 4C).
  • Storage is content dependent.
    Rabbit
    5 Plates
    78.13-5000 pg/mL
    B-cell antigen CD75, beta-galactoside alpha-2,6-sialyltransferase 1, CMP-N-acetylneuraminate beta-galactosamide alpha-2,6-sialyltransferase, CMP-N-acetylneuraminate-beta-galactosamide-alpha-2,6-sialyltransferase 1, EC 2.4.99.1, MGC48859, Sialyltransferase 1, sialyltransferase 1 (beta-galactoside alpha-2,6-sialyltransferase), sialyltransferase 1 (beta-galactoside alpha-2,6-sialytransferase), SIAT1Alpha 2,6-ST 1, ST6 beta-galactosamide alpha-2,6-sialyltranferase 1, ST6Gal I, ST6GalI, ST6N
    Conjugated
    SDS
    Documents
    Product Certifications
    Promotions

    Promotions

    Provide Content Correction

    We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Cancel Submit