Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ST6 Sialyltransferase 6/ST6GALNAC6 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP309229100UL
Description
ST6 Sialyltransferase 6/ST6GALNAC6 Polyclonal specifically detects ST6 Sialyltransferase 6/ST6GALNAC6 in Human samples. It is validated for Western Blot.Specifications
ST6 Sialyltransferase 6/ST6GALNAC6 | |
Polyclonal | |
Western Blot 1.0 ug/ml | |
alpha-N-acetylgalactosaminide alpha-2,6-sialyltransferase 6, CMP-NeuAC:(beta)-N-acetylgalactosaminide (alpha)2,6-sialyltransferase member VI, EC 2.4.99, EC 2.4.99.-, EC 2.4.99.7, GalNAc alpha-2,6-sialyltransferase VI, hST6GalNAc VI, RP11-203J24.3, Sialyltransferase 7F, sialytransferase 7 ((alpha-N-acetylneuraminyl 2,3-betagalactosyl-1,3)-N-acetylgalactosaminide alpha-2,6-sialytransferase) F, SIAT7F, SIAT7-F, ST6 (alpha-N-acetyl-neuraminyl-2,3-beta-galactosyl-1,3)-N-acetylgalactosaminidealpha-2,6-sialyltransferase 6, ST6GalNAc VI, ST6GalNAcVI | |
The immunogen is a synthetic peptide directed towards the N terminal region of human ST6 Sialyltransferase 6/ST6GALNAC6 (NP_038471). Peptide sequence ALITILILYSSNSANEVFHYGSLRGRSRRPVNLKKWSITDGYVPILGNKT | |
100 μg | |
Primary | |
Human | |
Purified |
Western Blot | |
Unconjugated | |
PBS buffer, 2% sucrose | |
Rabbit | |
Affinity purified | |
RUO | |
30815 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction