Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ST6GALNAC1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech Non-distribution product as customer accommodation.

Rabbit Polyclonal Antibody

Manufacturer:  Proteintech Group IncSupplier Diversity Partner 153631AP

Catalog No. 50-173-0179


Add to cart

Description

Description

Glycosylation of proteins affects cell-cell interaction, interactions with the matrix, and the functions of intracellular molecules. ST6GALNAC1 transfers a sialic acid, N-acetylneuraminic acid.
Specifications

Specifications

ST6GALNAC1
0.13 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q9NSC7, Q9QZ39
HSY11339, Sialyltransferase 7A, SIAT7 A, SIAT7A, ST6GalNAc I, ST6GALNAC1, ST6GalNAcI, STYI
Rabbit
IgG
150 μL
-20°C
Primary
20445, 55808
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
ST6GALNAC1
Liquid
ST6GALNAC1
ST6GALNAC1 Fusion Protein Ag8062
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit