Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ST8 alpha-2,8-Sialyltransferase 4/ST8SIA4 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP325159
Description
ST8 alpha-2,8-Sialyltransferase 4/ST8SIA4 Polyclonal antibody specifically detects ST8 alpha-2,8-Sialyltransferase 4/ST8SIA4 in Human samples. It is validated for Immunocytochemistry/ ImmunofluorescenceSpecifications
ST8 alpha-2,8-Sialyltransferase 4/ST8SIA4 | |
Polyclonal | |
Immunocytochemistry/Immunofluorescence 0.25 to 2 μg/mL | |
Alpha-2,8-sialyltransferase 8D, CMP-N-acetylneuraminate-poly-alpha-2,8-sialyl transferase, CMP-N-acetylneuraminate-poly-alpha-2,8-sialyltransferase, EC 2.4.99, EC 2.4.99.-, Polysialyltransferase-1, PST1MGC61459, PSTMGC34450, sialyltransferase 8 (alpha-2, 8-polysialytransferase) D, Sialyltransferase 8D, Sialytransferase St8Sia IV, SIAT8D, SIAT8-D, ST8 alpha-N-acetyl-neuraminide alpha-2,8-sialyltransferase 4, ST8SiaIV, ST8SIA-IV | |
This antibody has been engineered to specifically recognize the recombinant protein ST8 alpha-2,8-Sialyltransferase 4/ST8SIA4 using the following amino acid sequence: PSVVQRAFGGFRNESDREKFVHRLSMLNDSVLWIPAFMVKGGEKHVEWVNAL | |
100 μL | |
Neuroscience | |
7903 | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgG |
Immunocytochemistry | |
Unconjugated | |
PBS, pH 7.2, 40% glycerol | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction