Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ST8SIA6 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548789

Catalog No. PA548789


Add to cart

Description

Description

Prefers O-glycans to N-glycans or glycolipids as acceptor substrates. The minimal acceptor substrate is the NeuAc- alpha-2,3(6)-Gal sequence at the non-reducing end of their carbohydrate groups.
Specifications

Specifications

ST8SIA6
Polyclonal
PBS with 0.09% sodium azide
P61647
Alpha-2, 8-sialyltransferase 8F, 2499-, Sialyltransferase 8F, SIAT8-F, Sialyltransferase St8Sia VI, ST8SiaVI, ST8SIA6, SIAT8F
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
338596
Western Blot
Unconjugated
ST8SIA6
Liquid
ST8SIA6
KLH conjugated synthetic peptide between 138-167 amino acids from the Central region of human ST8SIA6.
Antigen Affinity Chromatography, Protein A
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit