Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Stabilin 1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA550834
Description
The antibody detects endogenous levels of total Stabilin 1 protein.
Stabilin 1 Polyclonal antibody specifically detects Stabilin 1 in Human samples. It is validated for Immunohistochemistry (Paraffin).Specifications
Stabilin 1 | |
Polyclonal | |
Unconjugated | |
STAB1 | |
CLEVER-1; common lymphatic endothelial and vascular endothelial receptor-1; fasciclin egf-like, laminin-type egf-like, and link domain-containing scavenger receptor-1; fasciclin, EGF-like, laminin-type EGF-like and link domain-containing scavenger receptor 1; FEEL1; FEEL-1; FEEL-1MS-1 antigen; FELE-1; FEX1; KIAA0246; MFEEL-1; mKIAA0246; MS-1; MS-1 antigen; SCARH2; scavenger receptor FEEL-1; STAB1; STAB-1; stabilin 1; stabilin-1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
23166 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
1 mg/mL | |
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
Q9NY15 | |
STAB1 | |
Synthetic peptide corresponding to residues near the C terminal of human stabilin 1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction