Learn More
Restek Stabilwax Capillary Columns
Low-bleed polyethylene glycol (PEG) column ensures long lifetimes
$372.00 - $2894.00
Specifications
Temperature Range | 40°C to 260°C |
---|---|
USP Type | G14, G15, G16, G20, G39 |
For Use With (Application) | FAMEs, Flavor Compounds, Essential Oils, Solvents, Aromatics, Acrolein/Acrylonitrile (EPA 603), and Oxygenated Compounds |
Polarity | Polar |
Stationary Phase | Crossbond Polyethylene Glycol (PEG) |
Catalog Number | Mfr. No. | Diameter (Metric) Inner | Film Thickness | Length (Metric) | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Diameter (Metric) Inner | Film Thickness | Length (Metric) | Price | Quantity & Availability | |||
06-610-751
|
Restek
42601 |
0.10 mm | 0.1 μm | 10 m |
Each for $495.00
|
|
06-610-752
|
Restek
43831 |
0.15 mm | 0.15 μm | 20 m |
Each for $578.00
|
|
06-610-731
|
Restek
10605 |
0.25 mm | 0.1 μm | 15 m |
Each for $384.00
|
|
06-610-732
|
Restek
10608 |
0.25 mm | 0.1 μm | 30 m |
Each for $578.00
|
|
06-610-733
|
Restek
10611 |
0.25 mm | 0.1 μm | 60 m |
Each for $1,008.00
|
|
06-610-736
|
Restek
10623600 |
0.25 mm | 0.25 μm | 30 m |
Pack of 6 for $2,894.00
|
|
06-610-737
|
Restek
10626 |
0.25 mm | 0.25 μm | 60 m |
Each for $1,022.00
|
|
06-610-740
|
Restek
10635 |
0.25 mm | 0.5 μm | 15 m |
Each for $372.00
|
|
06-610-743
|
Restek
10641 |
0.25 mm | 0.5 μm | 60 m |
Each for $1,008.00
|
|
06-610-734
|
Restek
10621 |
0.32 mm | 0.25 μm | 15 m |
Each for $389.00
|
|
06-610-738
|
Restek
10627 |
0.32 mm | 0.25 μm | 60 m |
Each for $1,095.00
|
|
06-610-741
|
Restek
10636 |
0.32 mm | 0.5 μm | 15 m |
Each for $394.00
|
|
06-610-745
|
Restek
10651 |
0.53 mm | 1 μm | 15 m |
Each for $389.00
|
|
06-610-735
|
Restek
10622 |
0.53 mm | 0.25 μm | 15 m |
Each for $415.00
|
|
Description
- Long column lifetimes due to the increased stability produced by the binding process
- Polar compound retention times do not shift
- Column can be rejuvenated by solvent washing
- Use for FAMEs, Flavor Compounds, Essential Oils, Solvents, Aromatics, Acrolein/Acrylonitrile (EPA 603), and Oxygenated Compounds
- Also used for purity testing of chemicals and analyzing alcoholic beverages
- Equivalent to USP G14, G15, G16, G20, and G39 phases
- Temperature range 40°C to 260°C
Specifications
40°C to 260°C | |
FAMEs, Flavor Compounds, Essential Oils, Solvents, Aromatics, Acrolein/Acrylonitrile (EPA 603), and Oxygenated Compounds | |
Crossbond Polyethylene Glycol (PEG) | |
Fused Silica |
G14, G15, G16, G20, G39 | |
Polar | |
Stabilwax™ |