Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
STAC2 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP309669100UL
Description
STAC2 Polyclonal specifically detects STAC2 in Human samples. It is validated for Western Blot, Immunoprecipitation.Specifications
STAC2 | |
Polyclonal | |
Western Blot 1.0 ug/ml, Immunoprecipitation | |
24b2/STAC2, MGC129694, SH3 and cysteine rich domain 2,24b2, SH3 and cysteine-rich domain-containing protein 2, Src homology 3 and cysteine-rich domain-containing protein 2 | |
The immunogen is a synthetic peptide directed towards the N-terminal region of Human STAC2 (NP_945344). Peptide sequence TEMSEKENEPDDAATHSPPGTVSALQETKLQRFKRSLSLKTILRSKSLEN | |
100 μg | |
Primary | |
Human | |
Purified |
Western Blot, Immunoprecipitation | |
Unconjugated | |
PBS buffer, 2% sucrose | |
Rabbit | |
Affinity purified | |
RUO | |
342667 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction