Learn More
Chemglass Life SciencesSupplier Diversity Partner Incubator, High Heat Decontamination, CO² (STACKING PLATFORM ONLY) DFS Item
Manufacturer:  Chemglass Life SciencesSupplier Diversity Partner CLS2447S56
Incubator, High Heat Decontamination, CO² (STACKING PLATFORM ONLY). Stacking platform for high heat decontamination incubator (item 50143490). Featuring a dry heat decontamination cycle, this model has the ability to maintain 180°C for 120 minutes. This time/temperature relationship satisfies all global standards for decontamination.Another important and unique feature is that it is not necessary to remove the IR CO² sensor prior to activating the decontamination process. This is a more user-friendly approach and eliminates potential damage to the sensitive IR sensor. This is decontamination at its easiest and most effective.Other features include a USB interface for software communication and a pre-heated copper CO² gas inlet. This exclusive feature prevents condensation and reduces potential sites for contamination to form. Precise temperature control is achieved via an advanced microprocessor. An independent over-temperature safety controller is also integral to the design.
Catalog No. 50-143-206