Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ STAG1 Monoclonal Antibody (GT8810)
GREENER_CHOICE

Mouse Monoclonal Antibody

Supplier:  Invitrogen™ MA527807

Catalog No. PIMA527807


Only null left
Add to Cart

Description

Description

Positive Control: 293T, A431, HeLa, HepG2 Predicted Reactivity: Mouse (100%), Rat (100%), Xenopus laevis (100%) Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

This gene is a member of the SCC3 family and is expressed in the nucleus. It encodes a component of cohesin, a multisubunit protein complex that provides sister chromatid cohesion along the length of a chromosome from DNA replication through prophase and prometaphase, after which it is dissociated in preparation for segregation during anaphase.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

STAG1
Monoclonal
1 mg/mL
PBS with no preservative
Q8WVM7
STAG1
Carrier-protein conjugated synthetic peptide encompassing a sequence within the C-terminus region of human SA1. The exact sequence is proprietary.
100 μL
Primary
Human, Rat
Antibody
IgG2a
Immunoprecipitation, Western Blot, Immunocytochemistry
GT8810
Unconjugated
STAG1
AU045003; cohesin subunit SA-1; DKFZp781D1416; nuclear protein stromal antigen 1; SA1; SA-1; SCC3; SCC3 homolog 1; SCC3A; STAG1; Stromal antigen 1
Mouse
Protein A
RUO
10274
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.