Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

STAG1 Rabbit anti-Human, Unconjugated, Polyclonal, Active Motif™ Non-distribution product as customer accommodation.

Rabbit Polyclonal Antibody

$135.30 - $534.00

Specifications

Monoclonal or Polyclonal Polyclonal
Primary or Secondary Primary
Antigen STAG1
Conjugate Unconjugated
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
50-199-3499 Non-distribution product as customer accommodation.
View Documents
Active MotifSupplier Diversity Partner
61562
10 μL
Each for $135.30
Add to cart
 
50-199-3498 Non-distribution product as customer accommodation.
View Documents
Active MotifSupplier Diversity Partner
61561
100 μL
Each for $534.00
Add to cart
 
Description

Description

This gene is a member of the SCC3 family and is expressed in the nucleus. It encodes a component of cohesin, a multisubunit protein complex that provides sister chromatid cohesion along the length of a chromosome from DNA replication through prophase and prometaphase, after which it is dissociated in preparation for segregation during anaphase.
Specifications

Specifications

Polyclonal
Primary
STAG1
Unconjugated
SDS
Product Certifications
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit