Learn More
STAG2 Mouse anti-Human, DyLight 350, Clone: 1417, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP243823UV
Catalog No. NBP243823UV
Add to cart
Description
Description
STAG2 Monoclonal antibody specifically detects STAG2 in Human samples. It is validated for Western Blot.
Specifications
Specifications
STAG2
1417
Western Blot
Purified
STAG2
Carrier-protein conjugated synthetic peptide encompassing a sequence within the C-terminus region of human SA2. The exact sequence is proprietary.
Protein A purified
DNA Repair
Primary
10735
Western Blot
DyLight 350
50mM Sodium Borate with 0.05% Sodium Azide
bA517O1.1, cohesin subunit SA-2, DKFZp686P168, SA-2, SA2FLJ25871, SCC3 homolog 2, stromal antigen 2DKFZp781H1753
Mouse
IgG2a
0.1 ml
Store at 4C in the dark.
Monoclonal
Human