Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

STAG2 Polyclonal Antibody

STAG2 Polyclonal Antibody

Supplier:  Thermo Scientific PA550784

Catalog No. PA550784


Only null left
Add to Cart

Description

Description

STAG2 Polyclonal Antibody for Western Blot, IHC (P)

TRUSTED_SUSTAINABILITY
Specifications

Specifications

STAG2
Polyclonal
Unconjugated
STAG2
9230105L23Rik; B230112I07Rik; bA517O1.1; cohesin subunit SA-2; DKFZp686P168; DKFZp781H1753; FLJ25871; nuclear protein SA2; OTTHUMP00000024338; OTTHUMP00000043514; OTTHUMP00000043515; OTTHUMP00000081754; OTTHUMP00000081755; RGD1562042; RP11-517O1.1; SA2; SA-2; Sap2; SCC3 homolog 2; SCC3B; STAG2; Stromal antigen 2
Rabbit
Antigen Affinity Chromatography
RUO
10735
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot
1.9 mg/mL
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4
Q8N3U4
STAG2
Fusion protein corresponding to residues near the C terminal of human stromal antigen 2.
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.