Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NHLRC1 Mouse anti-Human, Mouse, Rat, Clone: S85-18, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA527588

Catalog No. PIMA527588


Add to cart

Description

Description

STAG3 is a meiosis specific component of cohesin complex. The cohesin complex is required for the cohesion of sister chromatids after DNA replication. The cohesin complex apparently forms a large proteinaceous ring within which sister chromatids can be trapped. At anaphase, the complex is cleaved and dissociates from chromatin, allowing sister chromatids to segregate. The meiosis-specific cohesin complex probably replaces mitosis specific cohesin complex when it dissociates from chromatin during prophase I.
Specifications

Specifications

NHLRC1
Monoclonal
1 mg/ml
PBS with 50% glycerol and 0.1% sodium azide; pH 7.4
Q6IMG5, Q6VVB1, Q8BR37
E3 ubiquitin-protein ligase NHLRC1, NHLRC 1, NHL repeat containing 1, EPM2A, EPM2B, MGC119262, MGC119264, MGC119265, NHL repeat containing protein 1
Mouse
IgG1
100 μg
-20°C
Primary
105193, 364682, 378884
Immunocytochemistry, Immunofluorescence, Western Blot
S85-18
Unconjugated
NHLRC1
Liquid
NHLRC1
Fusion protein amino acids 2-125 (N-terminus encompassing RING domain) of human Malin. 86% identical to rat, and 77% identical to mouse.
Protein G
RUO
Antibody
Monoclonal
Human, Mouse, Rat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit