Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
STAG3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA549895
Description
The antibody detects endogenous levels of total STAG3 protein.
STAG3 Polyclonal antibody specifically detects STAG3 in Human samples. It is validated for Western Blot.Specifications
STAG3 | |
Polyclonal | |
Unconjugated | |
STAG3 | |
cohesin subunit SA-3; MGC119262; MGC119264; MGC119265; NHLRC 1; SA-2; SCC3 homolog 3; Stag3; Stromal antigen 3; stromalin 3; stromalin-3 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
10734 | |
-20°C | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q9UJ98 | |
STAG3 | |
Synthesized peptide derived from C-terminal of human STAG3. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction