Learn More
STAG3 Rabbit anti-Human, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 233141AP
Catalog No. 50-173-4046
Add to cart
Description
Description
STAG3 is a meiosis specific component of cohesin complex. The cohesin complex is required for the cohesion of sister chromatids after DNA replication. The cohesin complex apparently forms a large proteinaceous ring within which sister chromatids can be trapped. At anaphase, the complex is cleaved and dissociates from chromatin, allowing sister chromatids to segregate. The meiosis-specific cohesin complex probably replaces mitosis specific cohesin complex when it dissociates from chromatin during prophase I.
Specifications
Specifications
STAG3
0.31 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q9UJ98
Cohesin subunit SA 3, SCC3 homolog 3, STAG3, stromal antigen 3, Stromalin 3
Rabbit
IgG
150 μL
-20°C
Primary
10734
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation
Unconjugated
STAG3
Liquid
STAG3
STAG3 Fusion Protein Ag19818
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human