Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

STAG3L1 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA549142

Catalog No. PA549142


Add to cart

Specifications

Specifications

STAG3L1
Polyclonal
PBS with 0.09% sodium azide
P0CL83
Putative STAG3-like protein 1, Stromal antigen 3-like protein 1, STAG3L1
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
54441
Western Blot
Unconjugated
STAG3L1
Liquid
STAG3L1
KLH conjugated synthetic peptide between 108-136 amino acids from the Central region of human STAG3L1.
Protein A, Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit