Learn More
Cole-Parmer Meiji Techno MA286 Microscope Stage Micrometer, 0.04"/0.001" scale  DFS Item
Manufacturer:  Cole-Parmer 4840481
Use these micrometers to make quick and accurate measurements of your specimen. Eyepiece reticles are available with a crosshair scale or a 10 mm scale in 0.1 mm divisions; stage micrometers are available with a 1 mm scale in 0.01 mm divisions or with a 0.04" scale in 0.001" divisions. Use stage micrometers with any microscope.
Catalog No. 50-631-626