Learn More
MilliporeSigma™ Stage-Specific Embryonic Antigen-1, Clone: MC-480,™ (Chemicon™)
Supplier: MilliporeSigma™ MAB4301/DEL
Description
SSEA-1 is expressed on the surface of early mouse embryos, murine embryonal carcinoma cells (EC), murine embryonic stem cells (ES) and murine & human germ cells (EG). No immunoreactivity is evident with undifferentiated human EC and ES cells.Expression of SSEA-1 is down regulated following differentiation of murine EC and ES cells. In contrast, the differentiation of human EC and ES cells is characterized by an increase in SSEA-1 expression.
Vassilieva, S. et al(2000). Establishment of SSEA-1 and Oct-4 expressing Rat embryonic Stem-like cell lines and effects of cytokines of the IL-6 family on clonal growth.Exp. Cell Res 258: 361-373
Flow Cytometry, Immunocytochemistry, Immunofluorescence

Specifications
Stage-Specific Embryonic Antigen-1 | |
1mg/mL | |
Mouse | |
100 μg | |
Stem Cell Biology | |
Human, Mouse, Rat | |
IgM |
Monoclonal | |
Unconjugated | |
F9 teratocarcinoma stem cells (X-irradiated) | |
RUO | |
Primary | |
Purified |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.