Learn More
MilliporeSigma™ Supelco™ Stainless Steel 1/16 in. Capillary Tubing, Length: 20 cm
Use to connect the column to the detector compartment in a capillary gas chromatographic setup
$9.12 - $18.56
Specifications
Overall Length (Metric) 20 cm
Diameter (English) Outer 1/16 in.
Material Stainless Steel
Type Capillary Tubing
For Use With (Application) Gas Chromatography (GC)
View More Specs
Products 6
Catalog Number Mfr. No. Diameter (English) Inner Price Quantity & Availability  
Catalog Number Mfr. No. Diameter (English) Inner Price Quantity & Availability  
11-100-5275
View Documents
MilliporeSigma™ Supelco™
56709
0.005 in.
Each for $10.42
Add to cart
11-100-5279
View Documents
MilliporeSigma™ Supelco™
56715U
0.007 in.
Each for $9.76
Add to cart
11-100-5271
View Documents
MilliporeSigma™ Supelco™
56721
0.010 in.
Each for $11.29
Add to cart
11-100-5281
View Documents
MilliporeSigma™ Supelco™
56727
0.020 in.
Each for $9.12
Add to cart
11-100-5280
View Documents
MilliporeSigma™ Supelco™
56733U
0.030 in.
Each for $9.76
Add to cart
11-100-5237
View Documents
MilliporeSigma™ Supelco™
56739
0.040 in.
Each for $18.56
Add to cart
Specifications
Specifications
20 cm
Stainless Steel
Gas Chromatography (GC)
1/16 in.
Capillary Tubing