Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Supelco™ Stainless Steel 1/16 in. Capillary Tubing, Length: 5 cm

Use to connect the column to the detector compartment in a capillary gas chromatographic setup

$5.10 - $7.27

Specifications

Overall Length (Metric) 5 cm
Diameter (English) Outer 1/16 in.
Material Stainless Steel
Type Capillary Tubing
For Use With (Application) Gas Chromatography (GC)
View More Specs

Products 5
Catalog Number Mfr. No. Diameter (English) Inner Price Quantity & Availability  
Catalog Number Mfr. No. Diameter (English) Inner Price Quantity & Availability  
11-100-5297
View Documents
MilliporeSigma™ Supelco™
56707
0.005 in. Each for $5.21
Add to cart
 
11-100-5300
View Documents
MilliporeSigma™ Supelco™
56713
0.007 in. Each for $5.10
Add to cart
 
11-100-5301
View Documents
MilliporeSigma™ Supelco™
56719
0.010 in. Each for $5.10
Add to cart
 
11-100-5302
View Documents
MilliporeSigma™ Supelco™
56731
0.030 in. Each for $5.10
Add to cart
 
11-100-5287
View Documents
MilliporeSigma™ Supelco™
56737
0.040 in. Each for $7.27
Add to cart
 
Specifications

Specifications

5 cm
Stainless Steel
Gas Chromatography (GC)
1/16 in.
Capillary Tubing
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit