Alfa Aesar™ Stainless Steel flake, -150 mesh, 1.2 micron thick, Type 316-L

Manufacturer: Alfa Aesar™ 4248630

Quantity 250g

 View more versions of this product

Catalog No. AA4248630

  Check Availability
Add to Cart

    Chemical Identifiers

    MDL Number MFCD00148499
    Chemical Name or Material Stainless Steel flake, Type 316-L, 1.2 micron thick

    Specifications

    Quantity 250g
    Physical Form Flakes
    MDL Number MFCD00148499
    Mesh Size −150 Mesh
    TSCA TSCA
    Avg. Mol. Wt. or Mol. Wt. Range Fe:Cr:Ni:Mo; 67.5:17:13:2.5 wt%
    Chemical Name or Material Stainless Steel flake, Type 316-L, 1.2 micron thick

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit