Learn More
Alfa Aesar™ Stainless Steel flake, -325 mesh, 0.4-1.2 micron thick, Type 316-L
Manufacturer:  Alfa Aesar™ 13787A1
Quantity 1kg
 View more versions of this product
Catalog No. AA13787A1
  Check Availability
Add to cart
Specifications Description & Specifications
Chemical Identifiers
MFCD00148499
Stainless Steel flake, Type 316-L
Specifications
Fe:Cr:Ni:Mo; 67.5:17:13:2.5 wt%
1kg
Flakes
Stainless Steel flake, Type 316-L
−325 Mesh
0.4 to 1.2 Micron thick
MFCD00148499
Safety and Handling
SDS & Safety Documents
TSCA : TSCA