Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Stainless Steel Foil, 0.025mm (0.001in) thick, Alfa Aesar™

Manufacturer:  Alfa Aesar 41580FI

Quantity 50 x 50mm

 View more versions of this product

Catalog No. AA41580FI


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

Foil, 0.025mm (0.001 in.) thick
Fe:Cr:Ni; 70:19:11 wt%
MFCD00148499
Stainless Steel, Type 304

Specifications

Silver
Silver -colored foil
Foil, 0.025mm (0.001 in.) thick
MFCD00148499
50 x 50mm
Odorless
Fe:Cr:Ni; 70:19:11 wt%
Stainless Steel, Type 304
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit