Alfa Aesar™ Stainless Steel gauze, 100 mesh woven from 0.11mm (0.0045 in.) dia. wire, Type 316
Manufacturer: Alfa Aesar™ 46703FL
Quantity 75 x 75mm
 View more versions of this product
Catalog No. AA46703FL
  Check Availability
Add to Cart
Related Products
Product Content Feedback