Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Stainless Steel gauze, 325 mesh woven from 0.036mm (0.0014in) dia wire, Type 316, Thermo Scientific™

$23.86 - $81.32

Specifications

Chemical Name or Material Stainless Steel gauze, Type 316
Name Note 325 mesh woven from 0.036mm (0.0014 in.) dia. wire
MDL Number MFCD00148499
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
AA46715FL
View Documents
Thermo Scientific Chemicals
46715FL
75 x 75 mm
Each for $23.86
Order in Bulk

Request bulk or custom formats

Add to cart
 
AA46715GT
View Documents
Thermo Scientific Chemicals
46715GT
150 x 150 mm
Each for $40.54
Order in Bulk

Request bulk or custom formats

Add to cart
 
AA46715HB
View Documents
Thermo Scientific Chemicals
46715HB
300 x 300 mm
Each for $81.32
Order in Bulk

Request bulk or custom formats

Add to cart
 
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

Stainless Steel gauze, Type 316
325 mesh woven from 0.036mm (0.0014 in.) dia. wire
MFCD00148499
Safety and Handling

Safety and Handling

GHS H Statement
H372-H351-H317
Causes damage to organs through prolonged or repeated exposure.
Suspected of causing cancer.
May cause an allergic skin reaction.

P201-P202-P260-P264b-P270-P272-P280g-P281-P302+P352-P308+P313-P333+P313-P363-P501c

H317-H350-H372

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Product Certifications

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit