Alfa Aesar™ Stainless Steel gauze, 400 mesh woven from 0.028mm (0.0011 in.) dia. wire, twilled weave, Type 304

Manufacturer: Alfa Aesar™ 42012VA

Quantity 15 x 15cm

 View more versions of this product

Catalog No. AA42012VA

  Check Availability
Add to Cart

    Chemical Identifiers

    MDL Number MFCD00148499
    Chemical Name or Material Stainless Steel gauze, Type 304, Woven from 0.028mm (0.0011 in.) dia. wire

    Specifications

    Physical Form Twilled Weave Wire Cloth
    Quantity 15 x 15cm
    Health Hazard 3 GHS P Statement
    P260-P261-P280-P363-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    Wash contaminated clothing before reuse.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Odor Odorless
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H372-H351-H317
    Causes damage to organs through prolonged or repeated exposure.
    Suspected of causing cancer.
    May cause an allergic skin reaction.
    Mesh Size 400 mesh
    TSCA TSCA
    MDL Number MFCD00148499
    Avg. Mol. Wt. or Mol. Wt. Range Fe:Cr:Ni; 70:19:11 wt%
    Chemical Name or Material Stainless Steel gauze, Type 304, Woven from 0.028mm (0.0011 in.) dia. wire

    GHS H Statement
    H372-H351-H317
    Causes damage to organs through prolonged or repeated exposure.
    Suspected of causing cancer.
    May cause an allergic skin reaction.

    GHS P Statement
    P260-P261-P280-P363-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    Wash contaminated clothing before reuse.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit