Alfa Aesar™ Stainless Steel gauze, 80 mesh woven from 0.127mm (0.005 in.) dia. wire, Type 304

Manufacturer: Alfa Aesar™ 13477RU

Quantity 30 x 60cm

 View more versions of this product

Catalog No. AA13477RU

  Check Availability
Add to Cart

    Chemical Identifiers

    MDL Number MFCD00148499
    Chemical Name or Material Stainless Steel gauze, Type 304, Woven from 0.127mm (0.005 in.) dia. wire

    Specifications

    Quantity 30 x 60cm
    TSCA TSCA
    Physical Form Wire Cloth
    MDL Number MFCD00148499
    Mesh Size 80 mesh
    Avg. Mol. Wt. or Mol. Wt. Range Fe:Cr:Ni; 70:19:11 wt%
    Chemical Name or Material Stainless Steel gauze, Type 304, Woven from 0.127mm (0.005 in.) dia. wire

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit