Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Stainless-Steel gauze, Type 304, Alfa Aesar™

Manufacturer:  Alfa Aesar 13476RR

Quantity 10 x 10cm

 View more versions of this product

Catalog No. AA13476RR


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

Wire Cloth
Fe:Cr:Ni; 70:19:11 wt.%
MFCD00148499
Stainless Steel gauze, Open area: 36%; Width of opening: 0.003 in.

Specifications

10 x 10cm
Fe:Cr:Ni; 70:19:11 wt.%
Stainless Steel gauze, Open area: 36%; Width of opening: 0.003 in.
Wire Cloth
MFCD00148499

Safety and Handling

H317-H351: May cause an allergic skin reaction. Suspected of causing cancer.

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit